VeriSpec™ Nickel (Ni) Standard for AAS 30 ppb in 1% HNO₃, RICCA Chemical Company
Supplier: Ricca Chemical
RV010125-50N
RCRV010125-50NEA 239 USD
RCRV010125-50N
VeriSpec™ Nickel (Ni) Standard for AAS 30 ppb in 1% HNO₃, RICCA Chemical Company
Standards Cationic Standards Nickel Standards
Certified reference material (CRM) for AA (atomic absorption)
Certifications: Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility