VeriSpec® Sulfur (S) Standard for AAS 1000 ppm in H₂O, RICCA Chemical Company
Supplier: Ricca Chemical
RV010046-100N RV010046-500N
RCRV010046100NEA 70 USD
RCRV010046100N RCRV010046500N
VeriSpec® Sulfur (S) Standard for AAS 1000 ppm in H₂O, RICCA Chemical Company
Standards Cationic Standards Sulfur standards
Certified reference material (CRM) for AA (atomic absorption)
Certifications: Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility