Acetone for HPLC

Supplier: Ricca Chemical
Danger

Synonyms: 2-Propanone, DMK, Propanone, Dimethyl ketone

RSOA0011-4C
RCRSOA0011-4CEA 201 USD
RCRSOA0011-4C
Acetone for HPLC
Acetone
Formula: CH₃COCH₃
MW: 58.08 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00008765
CAS Number: 67-64-1
UN: 1090
ADR: 3,II

Order Now

Specification Test Results

Assay min 99.5 %
Color (APHA) max 10
Solubility in Water To Pass Test
Isopropyl Alcohol max 0.05 %
Methanol (Methyl Alcohol) max 0.05 %
Water (H₂O) max 0.5 %
Aldehyde (as HCHO) max 0.002 %
Residue after evaporation (Non-Volatile Matter) max 0.001 %
Substances Reducing Permanganate To Pass Test
Titratable Acid max 0.0003 meq/g
Titratable Base max 0.0006 meq/g
Absorbance at 330 nm 0.00-1.00
Absorbance at 340 nm 0.000-0.100
Absorbance at 350 nm 0.000-0.020
Absorbance at 400 nm 0.000-0.010

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR