Amidosulfonic acid, powder ACS

Supplier: Ricca Chemical
Danger

Synonyms: Aminosulphonic acid, Amidosulphuric acid, Sulfamic acid, Sulfamidic acid, Amidosulphuric Acid, Sulphamic acid, Amidosulphonic acid, Sulphamidic acid, Aminosulfonic acid, Amidosulfonic acid, Amidosulfuric Acid, Amidosulfuric acid

RDCS1100-500B1 RDCS1100-100B1 RDCS1100-2.5A5
RCRDCS1100500BEA 276 USD
RCRDCS1100500B RCRDCS110100B1 RCRDCS1102.5A5
Amidosulfonic acid, powder ACS
Amidosulphonic acid
Formula: H₃NO₃S
MW: 97.09 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011603
CAS Number: 5329-14-6
UN: 2967
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=afa0500786be69d531cdf06f8e67548e6f67b23f&vid=73aa20196fbabf965e4375f2cf3675e2f513ee59&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (dried basis) 99.3-100.3 %
Heavy Metals (as Pb) max 0.001 %
Sulfate (SO₄) max 0.05 %
Chloride (Cl) max 0.001 %
Iron (Fe) max 5 ppm
Insoluble Matter max 0.01 %
Residue after Ignition max 0.01 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR