Nitric acid ≥70% (w/w) for trace metal analysis

Supplier: Ricca Chemical
Danger

RABN0020-2.5D1
RCRABN00202.2DEA 531.99 USD
RCRABN00202.2D RCRABN002025D1
Nitric acid ≥70% (w/w) for trace metal analysis
Nitric acid
Formula: HNO₃
Storage Temperature: Ambient
MDL Number: MFCD00011349
CAS Number: 7697-37-2
UN: 2031
ADR: 8 (5.1),II

Order Now

Specification Test Results

Assay 68.0-70.0 % (w/w)
Lithium (Li) max 0.1 ppm
Boron (B) max 0.1 ppm
Sodium (Na) max 0.1 ppm
Magnesium (Mg) max 0.1 ppm
Aluminum (Al) max 0.1 ppm
Potassium (K) max 0.1 ppm
Calcium (Ca) max 0.1 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR