Zinc sulfate 1.00 M

Supplier: Ricca Chemical
Warning

R9665000-500A
RCR9665000500AEA 77 USD
RCR9665000500A
Zinc sulfate 1.00 M
Zinc sulphate
Formula: O₄SZn
MW: 161.45 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011302
CAS Number: 7733-02-0

Order Now

Specification Test Results

Appearance Colorless liquid
Assay (vs. EDTA/Xylenol Orange) 0.999-1.001 Molar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR