Zinc sulfate 0.0200 M (M/50)

Supplier: Ricca Chemical

R9657000-500A
RCR9657000500AEA 35 USD
RCR9657000500A
Zinc sulfate 0.0200 M (M/50)
Zinc sulphate
Formula: O₄SZn
MW: 161.45 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011302
CAS Number: 7733-02-0

Order Now

Specification Test Results

Appearance Colorless liquid
Assay (vs. EDTA/Xylenol Orange) 0.01998-0.02002 Molar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR