Water ASTM Type III

Supplier: Ricca Chemical

Synonyms: Dihydrogen oxide

R9151300-4A
RCR9151300-4AEA 148 USD
RCR9151300-4A
Water ASTM Type III
Water
Formula: H₂O
MW: 18.02 g/mol
Boiling Pt: 100 °C
Melting Pt: 0 °C
Density: 0.997 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011332
CAS Number: 7732-18-5

Order Now

Specification Test Results

Appearance Colorless liquid
Chloride (Cl) max 10 ppb
Conductivity at 25°C max 0.25 µS/cm
Organic Carbon (TOC) max 200 ppb
Silicate (as SiO₂) max 500 ppb
Sodium (Na) max 10 ppb

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR