Turbidity Standard, 18 NTU, Polymer Bead Suspension
Supplier: Ricca Chemical
R8818000-100C
RCR8818000100CEA 78 USD
RCR8818000100C
Turbidity Standard, 18 NTU, Polymer Bead Suspension
Standards Turbidity Standards
Traceable to commercially prepared EPA recognized primary standards.
  • Ready to use solution
Polymer bead suspension.