Trichloroacetic acid (TCA) 15% (w/v)

Supplier: Ricca Chemical
Danger

Synonyms: TCA, Trichloroethanoic acid, Aceto-Caustin, TCA

R8686500-4C
RCR8686500-4CEA 437 USD
RCR8686500-4C
Trichloroacetic acid (TCA) 15% (w/v)
Trichloroacetic acid (TCA)
Formula: Cl₃CCOOH
Storage Temperature: Ambient
MDL Number: MFCD00004177
CAS Number: 76-03-9
UN: 2564
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ae34130684af3f73eccad44eac686e325855232d&vid=b600bf2ba67ce275263e6172675e131d3f263837&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless liquid
Assay (vs. Sodium Hydroxide/Phenolphthalein) 14.95-15.05 % (w/v)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR