Total Ionic Strength Adjustment Buffer (TISAB II), with CDTA for Fluoride Analysis using Ion Selective Electrodes, Ricca Chemical Company

Supplier: Ricca Chemical

R8670000-10F
RCR8670000-10FEA 777 USD
RCR8670000-10F
Total Ionic Strength Adjustment Buffer (TISAB II), with CDTA for Fluoride Analysis using Ion Selective Electrodes, Ricca Chemical Company
Buffers Ionic Strength Adjustment Buffers

Specification
Appearance: Colorless liquid
pH at 25°C (Method: SQCP027, SQCP033): 5.30-5.50
Suitability for use: Fluoride analysis

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR