Toluidine blue Penetrant Solution ASTM F1929

Supplier: Ricca Chemical

Synonyms: Toluidine blue O zinc chloride double salt

R8656000-1A
RCR8656000-1AEA 187 USD
RCR8656000-1A
Toluidine blue Penetrant Solution ASTM F1929
Toluidine blue
Formula: C₁₅H₁₆ClN₃S·0.5ZnCl₂
MW: 747.96
Storage Temperature: Ambient
MDL Number: MFCD03548293
CAS Number: 6586-04-5

Order Now

Specification Test Results

Appearance Blue-purple liquid
UV/Visible Spectrum To Pass Test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR