Titration solvent ASTM D-2896 for total base number
Supplier: Ricca Chemical
R8621000-4C
RCR8621000-4CEA 1116 USD
RCR8621000-4C
Titration solvent ASTM D-2896 for total base number
Titration solvent
Chlorobenzene : acetic acid mixture