L(+)-Tartaric acid 10% (w/v)
Supplier: Ricca Chemical
|
Danger
|
Synonyms:
L-Tartaric Acid, (2R,3R)-(+)-Tartaric acid, Tartaric Acid, (R,R)-(+)-2,3-Dihydroxysuccinic acid, (R,R)-(+)-Tartaric acid, (2R,3R)-2,3-Dihydroxysuccinic acid
R8371000-1A
RCR8371000-1AEA
113
USD
RCR8371000-1A
L(+)-Tartaric acid 10% (w/v)
L(+)-Tartaric acid
Formula:
C₄H₆O₆ MW: 150.09 g/mol Boiling Pt: 399 Flash Pt: 150 Storage Temperature: Ambient |
MDL Number:
MFCD00064207 CAS Number: 87-69-4 Merck Index: 13,09158 |
|
Specification Test Results
Appearance | Colorless to slightly yellow liquid |
Assay (by Refractive Index) | 9.5-10.5 % (w/v) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...