Sulfuric acid 11.0 N

Supplier: Ricca Chemical
Danger

R8341000-1A
RCR8341000-1AEA 146 USD
RCR8341000-1A
Sulfuric acid 11.0 N
Sulfuric acid
Formula: H₂SO₄
MW: 98.08
Boiling Pt: 330
Melting Pt: 10.38
Density: 1.84
Storage Temperature: Ambient
MDL Number: MFCD00064589
CAS Number: 7664-93-9
UN: 2796
ADR: 8 II

Order Now

Specification Test Results

Appearance Colorless liquid
Assay (vs. Sodium Hydroxide/Phenolphthalein) 10.95-11.05 N at 20°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR