Sodium tetraphenylborate 0.02 M

Supplier: Ricca Chemical

Synonyms: Tetraphenyl sodium borate, Sodium tetraphenylboron, Tetraphenylboron sodium salt

R7855000-1A
RCR7855000-1AEA 132 USD
RCR7855000-1A
Sodium tetraphenylborate 0.02 M
Sodium tetraphenylborate
Formula: (C₆H₅)₄BNa
Storage Temperature: Ambient
MDL Number: MFCD00011494
CAS Number: 143-66-8

Order Now

Specification Test Results

Appearance Colorless to tan liquid
Assay (Calculated) 0.0198-0.0202 N at 20°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR