Sodium hydroxide 6.25 Normal

Supplier: Ricca Chemical
Danger

Synonyms: Caustic soda, Caustic soda, Sodium hydrate, Lye, Soda lye

R7467000-20F R7467000-4A
RCR7467000-20FEA 265 USD
RCR7467000-20F RCR7467000-4A
Sodium hydroxide 6.25 Normal
Sodium hydroxide
Formula: NaOH
Melting Pt: ∼ 0°C
Density: 1.23
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
UN: 1824
ADR: 8,II

Order Now

Specification Test Results

Appearance Colorless liquid
Assay (vs. Sulfuric Acid/Phenolphthalein) 6.245-6.255 N at 20°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR