Sodium hydroxide 1.00 Normal , from NF/USP/EP material, Tightrant™ Tested in an ISO 17025 Accredited Facility

Supplier: Ricca Chemical
Danger

Synonyms: Caustic soda, Caustic soda, Sodium hydrate, Lye, Soda lye

R7450010-1A R7450010-120A
RCR7450010-1AEA 107 USD
RCR7450010-1A RCR7450010120A
Sodium hydroxide 1.00 Normal , from NF/USP/EP material, Tightrant™ Tested in an ISO 17025 Accredited Facility
Sodium hydroxide
Formula: NaOH
Melting Pt: ∼ 0°C
Density: 1.04
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
UN: 1824
ADR: 8,III

Order Now

Specification Test Results

Appearance Colorless liquid
Assay (vs. Potassium Hydrogen Phthalate/Phenolphthalein) 0.999-1.001 N at 20°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR