Sodium hydroxide 12,5 g/L 0.313 Normal, Tightrant™ suitable for crude fiber analysis of food and feed Tested in an ISO 17025 Accredited Facility

Supplier: Ricca Chemical
Danger

Synonyms: Caustic soda, Caustic soda, Sodium hydrate, Lye, Soda lye

R7375000-4A R7375000-20F R7375000-1A R7375000-500A
RC73751EA 1029 USD
RC73751 RC73755 RCR7375000-1A RCR7375000500A
Sodium hydroxide 12,5 g/L 0.313 Normal, Tightrant™ suitable for crude fiber analysis of food and feed Tested in an ISO 17025 Accredited Facility
Sodium hydroxide

AOAC, for crude fiber.

Formula: NaOH
Melting Pt: 0.0°C
Density: 1.01
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
UN: 1824
ADR: 8,III

Order Now

Specification Test Results

Appearance Colorless liquid
Assay (vs. Potassium Hydrogen Phthalate/Phenolphthalein) 0.3125-0.3135 N at 20°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR