Sodium hydroxide 0.100 Normal (N/10) , from NF/USP material, Tightrant™ Tested in an ISO 17025 Accredited Facility

Supplier: Ricca Chemical

Synonyms: Caustic soda, Caustic soda, Sodium hydrate, Lye, Soda lye

R7350005-1A R7350005-4A
RCR7350005-1AEA 119 USD
RCR7350005-1A RCR7350005-4A
Sodium hydroxide 0.100 Normal (N/10) , from NF/USP material, Tightrant™ Tested in an ISO 17025 Accredited Facility
Sodium hydroxide
Formula: NaOH
Melting Pt: 0.0°C
Density: 1
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2

Order Now

Specification Test Results

Appearance Colorless liquid
Assay (vs. Potassium Hydrogen Phthalate/Phenolphthalein) 0.0999-0.1001 N at 20°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR