Sodium hydroxide 50% (w/v)
Supplier: Ricca Chemical
|
Danger
|
Synonyms:
Caustic soda, Caustic soda, Sodium hydrate, Lye, Soda lye
R7282000-20F
R7282000-4A
RCR7282000-20FEA
1533
USD
RCR7282000-20F
RCR7282000-4A
Sodium hydroxide 50% (w/v)
Sodium hydroxide
Formula:
NaOH Boiling Pt: ~100 °C Melting Pt: ~0 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003548 CAS Number: 1310-73-2 EINECS: 215-185-5 UN: 1824 ADR: 8,II |
Specification Test Results
Appearance | Colorless liquid |
Assay (vs. Sulfuric Acid/Phenolphthalein) | 49.92 - 50.08 % (w/v) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...