Sodium hydroxide 20% (w/v) in aqueous solution (200 g/L) analytical reagent

Supplier: Ricca Chemical
Danger

Synonyms: Caustic soda, Caustic soda, Sodium hydrate, Lye, Soda lye

R7270000-4A R7270000-500A R7270000-1A R7271000-20F R7271000-4A
RC72701EA 175 USD
RC72701 RC727016 RC727032 RCR7271000-20F RCR7271000-4A
Sodium hydroxide 20% (w/v) in aqueous solution (200 g/L) analytical reagent
Sodium hydroxide

Certifications: P4TF8-S12, Class F

Formula: NaOH
Boiling Pt: 119…135 °C (1013 hPa)
Melting Pt: 9…10 °C
Density: 1.53 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: 1824
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear, colorless, odorless
Assay at 20 °C (traceable to NIST SRM 723) 20.00 ± 0.04 % (w/v)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR