Potassium carbonate 15% (w/v)

Supplier: Ricca Chemical

R5883150-500A
RCR5883150500AEA 221 USD
RCR5883150500A
Potassium carbonate 15% (w/v)
Potassium carbonate
Formula: K₂CO₃
Boiling Pt: ∼100 °C
MDL Number: MFCD00011382
CAS Number: 584-08-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4d5900faa1ba52c3213eed8f6bb2d952bac1f30f&vid=6298361525e6af416556b5694c74a8a91faa437c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless Liquid
Physical State Liquid
pH 12
Initial Boiling Point /Range Approximately 100 °C
Relative Density 1.13
Solubility Miscible
Acute Toxicity LD50, Oral, Rat: 1870 mg/kg (Potassium Carbonate), details of toxic effects not reported other than lethal dose value


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR