Potassium carbonate 3% (w/v)

Supplier: Ricca Chemical
Warning

R5882300-1A
RCR5882300-1AEA 222 USD
RCR5882300-1A
Potassium carbonate 3% (w/v)
Potassium carbonate
Formula: K₂CO₃
Boiling Pt: ~100 °C
Melting Pt: ~0 °C
Storage Temperature: Ambient
MDL Number: MFCD00011382
CAS Number: 584-08-7

Order Now

Specification Test Results

Appearance Colorless liquid
Assay (vs. Sulfuric Acid/Methyl Orange) 2.99 - 3.01 % (w/v)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR