Orthophosphoric acid 1.00 N

Supplier: Ricca Chemical

R5842500-500A
RCR5842500500AEA 311 USD
RCR5842500500A
Orthophosphoric acid 1.00 N
Orthophosphoric acid
Formula: H₃PO₄
MDL Number: MFCD00011340
CAS Number: 7664-38-2
UN: 1805
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR