Phosphate Buffered Saline, 0.01 Molar, pH 7.40 at 25°C, RICCA Chemical Company
Supplier: Ricca Chemical
R5823500-10F
RCR5823500-10FEA
437
USD
RCR5823500-10F
Phosphate Buffered Saline, 0.01 Molar, pH 7.40 at 25°C, RICCA Chemical Company
Buffers
Electrophoresis Buffers
Washing Buffers
Specifications
Appearance: Colorless liquid
Assay (vs. Silver Nitrate/Potassium Chromate): 0.89-0.91 % (w/v)
pH at 25°C (Method: SQCP027, SQCP033): 7.38-7.42
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...