Nitrite Standard, 1 ml = 1 mg N, 1000 ppm N as Nitrite (3285 ppm NO₂⁻), Ricca Chemical Company
Supplier: Ricca Chemical
R5444900-120C
RCR5444900120CEA 59 USD
RCR5444900120C
Nitrite Standard, 1 ml = 1 mg N, 1000 ppm N as Nitrite (3285 ppm NO₂⁻), Ricca Chemical Company
Standards Anionic Standards Nitrite Standards