Nitric acid ≥70% (v/v)

Supplier: Ricca Chemical
Danger

Synonyms: Nitric acid solution

R5354700-2.5D
RCR53547002.5DEA 251 USD
RCR53547002.5D
Nitric acid ≥70% (v/v)
Nitric acid
Formula: HNO₃
MW: 63.01 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011349
CAS Number: 7697-37-2
UN: 2031
ADR: 8,II

Order Now

Specification Test Results

Appearance Colorless to light yellow liquid
Assay (vs. Sodium Hydroxide/Phenolphthalein) 69.3-70.7 % (v/v)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR