Magnesium hydroxide in aqueous solution 80 g/L

Supplier: Ricca Chemical

R4485000-500A
RCR4485000500AEA 224 USD
RCR4485000500A
Magnesium hydroxide in aqueous solution 80 g/L
Magnesium hydroxide
Formula: Mg(OH)₂
MW: 58.32 g/mol
Boiling Pt: ~100 °C
Melting Pt: ~0 °C
Storage Temperature: Ambient
MDL Number: MFCD00011104
CAS Number: 1309-42-8

Order Now

Specification Test Results

Appearance Milky white suspension
Assay (vs. EDTA/Eriochrome Black T) 77 - 83 g/L

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR