Ionic Strength Adjustor for use with Sodium Ion Selective Electrode, RICCA Chemical Company
Supplier: Ricca Chemical
R4130000-4A
RCR4130000-4AEA 230 USD
RCR4130000-4A
Ionic Strength Adjustor for use with Sodium Ion Selective Electrode, RICCA Chemical Company
Buffers Ionic Strength Adjustment Buffers
Ionic Strength Adjustor, for use with Sodium Ion Selective Electrode, packed in a natural poly Container
Colourless liquid.
Certifications: Certified suitable for use as an Ionic Strength Adjustor (ISA) for concentration measurement of Sodium Ion (Na+).