Hydrofluoric acid 0,1% w/w

Supplier: Ricca Chemical
Danger

R3815300-4A
RCR3815300-4AEA 301 USD
RCR3815300-4A
Hydrofluoric acid 0,1% w/w
Hydrofluoric acid
Formula: HF
MW: 20.01
Storage Temperature: Ambient
MDL Number: MFCD00011346
CAS Number: 7664-39-3

Order Now

Specification Test Results

Appearance Colorless liquid
Assay (vs. Sodium Hydroxide/Phenolphthalein) 0.095-0.105 % (w/w)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR