Hydrochloric acid 12.0 N

Supplier: Ricca Chemical
Danger

R3780000-1A R3780000-2.5D
RCR3780000-1AEA 110 USD
RCR3780000-1A RCR37800002.5D
Hydrochloric acid 12.0 N
Hydrochloric acid
Formula: HCl
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
UN: 1789
ADR: 8,II

Order Now

Specification Test Results

Appearance Colorless to slightly greenish-yellow liquid
Assay (vs. Sodium Hydroxide/Phenolphthalein) 11.8-12.2 N at 20°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR