Benzethonium chloride solution 0.004 M (0.004 N)

Supplier: Ricca Chemical

Synonyms: Hyamine® 1622, N-Benzyl-N,N-dimethyl-2-(2-(4-(2,4,4-trimethylpentan-2-yl)phenoxy)ethoxy)ethanaminium chloride

R3549000-500C
RCR3549000500CEA 283 USD
RCR3549000500C
Benzethonium chloride solution 0.004 M (0.004 N)
Benzethonium chloride
Formula: C₂₇H₄₂ClNO₂
MW: 448.09 g/mol
MDL Number: MFCD00011742
CAS Number: 121-54-0
UN: 3077
ADR: 9,III

Order Now

Specification Test Results

Assay 0.004 ± 0.0004 M (N) at 20 °C
NIST SRM 999

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR