EDTA (ethylenediamine tetraacetic acid) 0.100 M (M/10) with Magnesium titrant

Supplier: Ricca Chemical

R2729000-1A
RCR2729000-1AEA 68 USD
RCR2729000-1A
EDTA (ethylenediamine tetraacetic acid) 0.100 M (M/10) with Magnesium titrant
EDTA (Ethylenediamine tetraacetic acid)
Formula: C₁₀H₁₆N₂O₈
Storage Temperature: Ambient
MDL Number: MFCD00003541
CAS Number: 60-00-4

Order Now

Specification Test Results

Appearance Colorless liquid
Assay (vs. Calcium Carbonate/Eriochrome Black T) 0.0999-0.1001 Molar


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR