Diphenylamine 1% (w/v) in sulfuric acid

Supplier: Ricca Chemical
Danger

Synonyms: N-Phenylaniline, N-Phenylbenzeneamine

R2584000-120C
RCR2584000120CEA 70 USD
RCR2584000120C
Diphenylamine 1% (w/v) in sulfuric acid
Diphenylamine
Formula: (C₆H₅)₂NH
MW: 169.23 g/mol
Boiling Pt: 302
Flash Pt: 153
Storage Temperature: Ambient
MDL Number: MFCD00003014
CAS Number: 122-39-4
UN: 1830
ADR: 8 II
Merck Index: 13,03349

Order Now

Specification Test Results

Appearance Colorless liquid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR