Conductivity Standard, 200000 µS/cm (200 mS/cm) at 25 °C
Supplier: Ricca Chemical
R2249200-500A
RCR2249200500AEA 51 USD
RCR2249200500A
Conductivity Standard, 200000 µS/cm (200 mS/cm) at 25 °C
Standards Conductivity Standards
  • Certified traceable to NIST Standard Reference Materials
  • Tested in ISO 17025 Accredited Laboratories
Calibrate with confidence using RICCA Conductivity Standards. Suitalbe for use with any probe. Tested to the tightest specifications in the industry, you can count on RICCA conductivity buffers to produce accurate and reproducible readings with every pour.