Calcium Standard, 500 ppm Ca
Supplier: Ricca Chemical
R1799000-500A
RCR1799000500AEA
129
USD
RCR1799000500A
Calcium Standard, 500 ppm Ca
Standards
Cationic Standards
Calcium Standards
500 ppm Calcium standard for analysis.
- Single element standard
Ideal for instrument calibration.
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)