Ammonium sulfide USP Test Solution (TS)

Supplier: Ricca Chemical
Danger

R0694600-120C
RCR0694600120CEA 200 USD
RCR0694600120C
Ammonium sulfide USP Test Solution (TS)
Ammonium sulphide
Formula: H₈N₂S
MW: 68.14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00010892
CAS Number: 12135-76-1
UN: 2683
ADR: 8 (6.1,3),II

Order Now

Specification Test Results

Appearance Yellow liquid
Suitability for Use No turbidity with Magnesium Sulfate TS
Residue after Ignition max 0.05 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR