Ammonia solution 1% (w/w)

Supplier: Ricca Chemical
Warning

Synonyms: Ammonia aqueous, Ammonium hydroxide solution, Ammonium hydroxide, Ammonia water

R0631100-4A
RCR0631100-4AEA 146 USD
RCR0631100-4A
Ammonia solution 1% (w/w)
Ammonia solution
Formula: NH₃
MW: 35.05 g/mol
Boiling Pt: 100 °C
Melting Pt: 0.0 °C
Storage Temperature: Ambient
MDL Number: MFCD00066650
CAS Number: 1336-21-6

Order Now

Specification Test Results

Appearance Colorless liquid
Assay (vs. Sulfuric Acid/Methyl Red) 0.96 - 1.04 % (w/w)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR