Tellurium ICP Standard, 1 ml = 10 mg Te (10000 ppm Te)Te in 25% HCl, Ricca Chemical Company
Supplier: Ricca Chemical
RPTE10KH-100N
RCPTE10KH-100EA 228 USD
RCPTE10KH-100
Tellurium ICP Standard, 1 ml = 10 mg Te (10000 ppm Te)Te in 25% HCl, Ricca Chemical Company
Standards Cationic Standards Tellurium Standards