Tantalum ICP Standard, 1 ml = 1 mg Ta (1000 ppm Ta)(NH₄)₂TaF₇ in H₂O / trace HF, Ricca Chemical Company
Supplier: Ricca Chemical
RPTA1KW0-100N
RCPTA1KW-100EA 134 USD
RCPTA1KW-100
Tantalum ICP Standard, 1 ml = 1 mg Ta (1000 ppm Ta)(NH₄)₂TaF₇ in H₂O / trace HF, Ricca Chemical Company
Standards Cationic Standards Tantalum Standards