Sulfur ICP Standard, 1 ml = 10 mg S (10000 ppm S)(NH₄)2SO₄ in H₂O, Ricca Chemical Company
Supplier: Ricca Chemical
RPS10KW0-100N
RPS10KW0-50N
RPS10KW0-500N
RCPS10KW-100EA
559
USD
RCPS10KW-100
RCPS10KW-50
RCPS10KW-500
Sulfur ICP Standard, 1 ml = 10 mg S (10000 ppm S)(NH₄)2SO₄ in H₂O, Ricca Chemical Company
Standards
Cationic Standards
Sulfur standards