Neodymium ICP Standard, 1 ml = 10 mg Nd (10000 ppm Nd)Nd₂O₃ in 5% HNO₃, Ricca Chemical Company
Supplier: Ricca Chemical
RPND10KN-100N
RCPND10KN-100EA 244 USD
RCPND10KN-100
Neodymium ICP Standard, 1 ml = 10 mg Nd (10000 ppm Nd)Nd₂O₃ in 5% HNO₃, Ricca Chemical Company
Standards Cationic Standards Neodymium Standards