Mercury ICP Standard, 1 mL = 10 mg Hg (10000 ppm Hg)Hg in 10% HNO₃, Ricca Chemical Company
Supplier: Ricca Chemical
RPHG10KN-500N
RCPHG10KN-500EA
429
USD
RCPHG10KN-500
Mercury ICP Standard, 1 mL = 10 mg Hg (10000 ppm Hg)Hg in 10% HNO₃, Ricca Chemical Company
Standards
Cationic Standards
Mercury Standards
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)