Gallium ICP Standard, 1 ml = 1 mg Ga (1000 ppm Ga)Ga in 3% HNO₃, Ricca Chemical Company
Supplier: Ricca Chemical
RPGA1KN0-100N
RCPGA1KN-100EA 146 USD
RCPGA1KN-100
Gallium ICP Standard, 1 ml = 1 mg Ga (1000 ppm Ga)Ga in 3% HNO₃, Ricca Chemical Company
Standards Cationic Standards Gallium Standards