Scandium ICP-MS Standard, 1 ml = 1 mg Sc (1,000 ppm Sc)Sc₂O₃ in 3% HNO₃, Ricca Chemical Company
Supplier: Ricca Chemical
RMSSC1KN-100N
RCMSSC1KN-100EA 415 USD
RCMSSC1KN-100
Scandium ICP-MS Standard, 1 ml = 1 mg Sc (1,000 ppm Sc)Sc₂O₃ in 3% HNO₃, Ricca Chemical Company
Standards Cationic Standards Scandium Standards