Scandium ICP Standard, 1 ml = 0.01 mg Sc (10 ppm Sc)Sc₂O₃ in 3% HNO₃, Ricca Chemical Company
Supplier: Ricca Chemical
RMSSC10N-100N
RCMSSC10N-100EA 227 USD
RCMSSC10N-100
Scandium ICP Standard, 1 ml = 0.01 mg Sc (10 ppm Sc)Sc₂O₃ in 3% HNO₃, Ricca Chemical Company
Standards Cationic Standards Scandium Standards