Cobalt ICP-MS Standard, 1 ml = 1 mg Co (1000 ppm Co)Cobalt Metal in 3% HNO₃, Ricca Chemical Company
Supplier: Ricca Chemical
RMSCO1KN-100N
RCMSCO1KN-100EA
200
USD
RCMSCO1KN-100
Cobalt ICP-MS Standard, 1 ml = 1 mg Co (1000 ppm Co)Cobalt Metal in 3% HNO₃, Ricca Chemical Company
Standards
Cationic Standards
Cobalt Standards
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)