Gold ICP-MS Standard, 1 ml = 1 mg Au (1,000 ppm Au)Au in 3% HCl, Ricca
Supplier: Ricca Chemical
RMSAU1KH-100C
RCMSAU1KH-100EA 586 USD
RCMSAU1KH-100
Gold ICP-MS Standard, 1 ml = 1 mg Au (1,000 ppm Au)Au in 3% HCl, Ricca
Standards Cationic Standards Gold Standards