Lanthanum 5% in nitric acid 5% releasing agent

Supplier: Ricca Chemical

RMLA50KN-50N
RCMMLA50KN-50EA 693 USD
RCMMLA50KN-50
Lanthanum 5% in nitric acid 5% releasing agent
Lanthanum
Formula: La
MDL Number: MFCD00011066
CAS Number: 7439-91-0
UN: 3208
ADR: 4.3,II

Order Now

Specification Test Results

Appearance Clear, colorless liquid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR